Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MURE
Entrez Gene ID
842672
Gene Name
ALBINO OR PALE-GREEN 13
Gene Aliases
ALBINO OR PALE-GREEN 13, APG13, AT1G63680, ATMURE, F24D7.13, F24D7_13, PDE316, PIGMENT DEFECTIVE EMBRYO 316
Chromosome Location
Chr.1: 23614340 - 23617912 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334120.1 NP_001321889.1 2 - 3 2100 87
NM_001334121.1 NP_001321888.1 2 - 3 2056 87
NM_105045.6 NP_176555.4 2 - 3 2100 87

View Full Product Details