Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FLA2
Entrez Gene ID
826885
Gene Name
FASCICLIN-like arabinogalactan 2
Gene Aliases
AT4G12730, FASCICLIN-like arabinogalactan 2, T20K18.80, T20K18_80
Chromosome Location
Chr.4: 7491227 - 7493080 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117342.3 NP_193009.1 1 - 1 1626 115
GenBank mRNA AY060582.1 - 1 - 1 1407 115

View Full Product Details