Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl20
Entrez Gene ID
826903
Gene Name
serine carboxypeptidase-like 20
Gene Aliases
AT4G12910, F25G13.7, F25G13_7, serine carboxypeptidase-like 20
Chromosome Location
Chr.4: 7550238 - 7553375 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340813.1 NP_001329616.1 7 - 8 1100 83
NM_117360.8 NP_193027.5 7 - 8 1143 83

View Full Product Details