Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHF1A
Entrez Gene ID
827062
Gene Name
RING-H2 group F1A
Gene Aliases
AT4G14220, DL3150W, FCAALL.146, RING-H2 group F1A
Chromosome Location
Chr.4: 8195881 - 8198586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117499.5 NP_193158.2 6 - 7 928 100
GenBank mRNA DQ059122.1 - 6 - 7 797 100

View Full Product Details