Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APS3
Entrez Gene ID
827118
Gene Name
Pseudouridine synthase/archaeosine transglycosylase-like family protein
Gene Aliases
AT4G14680, DL3380C, FCAALL.128
Chromosome Location
Chr.4: 8413263 - 8415650 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340955.1 NP_001329676.1 1 - 2 795 88
NM_117550.5 NP_193204.1 1 - 2 840 88

View Full Product Details