Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPR4
Entrez Gene ID
827155
Gene Name
membrane-associated progesterone binding protein 4
Gene Aliases
AT4G14965, AtMAPR4, membrane-associated progesterone binding protein 4
Chromosome Location
Chr.4: 8551244 - 8553618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340979.1 NP_001329526.1 1 - 2 215 149
NM_117583.8 NP_567451.1 1 - 2 215 149
GenBank mRNA AK229457.1 - 1 - 2 215 149
BT024469.1 - 1 - 2 103 149

View Full Product Details