Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARA1
- Entrez Gene ID
-
827299
- Gene Name
- arabinose kinase
- Gene Aliases
-
ARABINOSE KINASE, AT4G16130, ATISA1, DL4105W, FCAALL.288, ISA1, arabinose kinase
- Chromosome Location
-
Chr.4: 9120777 - 9127938 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117706.3 |
NP_193348.1 |
13 - 14 |
1332 |
63 |
View Full Product Details