Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCI17
Entrez Gene ID
843122
Gene Name
non-intrinsic ABC protein 3
Gene Aliases
ARABIDOPSIS THALIANA NON-INTRINSIC ABC PROTEIN 3, AT1G67940, ATNAP3, ATP-binding cassette I17, AtSTAR1, T23K23.21, T23K23_21, non-intrinsic ABC protein 3
Chromosome Location
Chr.1: 25477670 - 25478803 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105464.3 NP_176961.1 1 - 2 484 90
GenBank mRNA AB567722.1 - 1 - 2 349 90
AK117705.1 - 1 - 2 430 90
AY086511.1 - 1 - 2 484 90
BT005105.1 - 1 - 2 349 90

View Full Product Details