Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- APRR6
- Entrez Gene ID
-
843150
- Gene Name
- pseudo-response regulator 6
- Gene Aliases
-
AT1G68210, PRR6, T22E19.16, T22E19_16, pseudo-response regulator 6
- Chromosome Location
-
Chr.1: 25565983 - 25569302 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_105492.4 |
NP_176988.4 |
2 - 3 |
265 |
161 |
View Full Product Details