Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22C
Entrez Gene ID
843306
Gene Name
HVA22 homologue C
Gene Aliases
AT1G69700, ATHVA22C, HVA22 homologue C, T6C23.10, T6C23_10
Chromosome Location
Chr.1: 26220190 - 26221870 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105638.4 NP_177128.1 3 - 4 361 100
GenBank mRNA AF141661.1 - 3 - 4 286 100
AY065456.1 - 3 - 4 308 100
AY084473.1 - 3 - 4 313 100
AY142652.1 - 3 - 4 214 100

View Full Product Details