Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTP1
Entrez Gene ID
843516
Gene Name
protein tyrosine phosphatase 1
Gene Aliases
AT1G71860, ATPTP1, F14O23.24, F14O23_24, PROTEIN TYROSINE PHOSPHATASE 1, protein tyrosine phosphatase 1
Chromosome Location
Chr.1: 27026677 - 27029092 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036189.1 NP_001031266.1 7 - 8 1097 89
NM_105844.4 NP_177331.1 7 - 8 1131 89
NM_202398.1 NP_974127.1 7 - 8 1074 89
GenBank mRNA AY070403.1 - 7 - 8 1079 89
AY117240.1 - 7 - 8 942 89

View Full Product Details