Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGLU11
Entrez Gene ID
839435
Gene Name
beta glucosidase 11
Gene Aliases
AT1G02850, F22D16.15, F22D16_15, beta glucosidase 11
Chromosome Location
Chr.1: 630374 - 633420 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123745.2 NP_001117217.1 2 - 3 397 69
NM_001331360.1 NP_001318909.1 2 - 3 397 69
NM_100166.4 NP_563666.1 2 - 3 397 69
NM_179247.6 NP_849578.5 2 - 3 397 69
NM_202017.3 NP_973746.3 2 - 3 266 69
GenBank mRNA AY049274.1 - 2 - 3 258 69
AY062763.1 - 2 - 3 259 69
BT001137.1 - 2 - 3 202 69

View Full Product Details