Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIB2
Entrez Gene ID
828668
Gene Name
fibrillarin 2
Gene Aliases
AT4G25630, ATFIB2, L73G19.10, L73G19_10, fibrillarin 2
Chromosome Location
Chr.4: 13074090 - 13076432 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118695.4 NP_567724.1 6 - 7 927 87
GenBank mRNA AF233444.1 - 6 - 7 856 87
AY084608.1 - 6 - 7 849 87
AY142647.1 - 6 - 7 778 87

View Full Product Details