Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT-HF
Entrez Gene ID
828797
Gene Name
HIS HF
Gene Aliases
AT4G26900, HIS HF, HISN4
Chromosome Location
Chr.4: 13515432 - 13519867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118824.4 NP_194420.1 4 - 5 545 98

View Full Product Details