Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HST
Entrez Gene ID
5007993
Gene Name
homogentisate prenyltransferase
Gene Aliases
AT3G11945, ATHST, PDS2, PHYTOENE DESATURATION 2, homogentisate prenyltransferase
Chromosome Location
Chr.3: 3779839 - 3782981 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084669.2 NP_001078138.1 5 - 6 886 73
NM_001161137.1 NP_001154609.1 6 - 7 880 73
GenBank mRNA DQ231060.1 - 5 - 6 785 73

View Full Product Details