Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THI1
Entrez Gene ID
835567
Gene Name
thiazole biosynthetic enzyme, chloroplast (ARA6) (THI1) (THI4)
Gene Aliases
AT5G54770, MBG8.3, MBG8_3, THI1 PROTEIN, THI4, THIAMINE4, THIAZOLE REQUIRING, TZ
Chromosome Location
Chr.5: 22246519 - 22248332 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124858.4 NP_200288.1 1 - 2 516 56
GenBank mRNA AF419604.1 - 1 - 2 440 56
AF428355.1 - 1 - 2 440 56
AF428385.1 - 1 - 2 440 56
AY054216.1 - 1 - 2 440 56
AY058094.1 - 1 - 2 440 56
AY143082.1 - 1 - 2 401 56
U17589.1 - 1 - 2 422 56

View Full Product Details