Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAX2
Entrez Gene ID
820532
Gene Name
cation exchanger 2
Gene Aliases
AT3G13320, MDC11.19, atcax2, cation exchanger 2
Chromosome Location
Chr.3: 4314505 - 4318487 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112177.3 NP_566452.1 11 - 12 1690 127
GenBank mRNA AF424628.1 - 11 - 12 1375 127
AY097343.1 - 10 - 11 1226 127

View Full Product Details