Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OVA5
Entrez Gene ID
820551
Gene Name
Lysyl-tRNA synthetase, class II
Gene Aliases
ARABIDOPSIS THALIANA LYSYL-TRNA SYNTHETASE 2, AT3G13490, ATKRS-2, OVULE ABORTION 5
Chromosome Location
Chr.3: 4395663 - 4399350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112195.3 NP_187958.1 1 - 2 328 99
GenBank mRNA BT002317.1 - 1 - 2 280 99

View Full Product Details