Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABA1g
Entrez Gene ID
820735
Gene Name
RAB GTPase homolog A1G
Gene Aliases
AT3G15060, AtRABA1g, RAB GTPase homolog A1G
Chromosome Location
Chr.3: 5069104 - 5070540 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112368.4 NP_188124.1 1 - 2 357 102
GenBank mRNA AK118727.1 - 1 - 2 279 102
BT005565.1 - 1 - 2 222 102

View Full Product Details