Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAMMA-VPE
Entrez Gene ID
829431
Gene Name
gamma vacuolar processing enzyme
Gene Aliases
AT4G32940, F26P21.60, F26P21_60, GAMMAVPE, gamma vacuolar processing enzyme
Chromosome Location
Chr.4: 15900270 - 15903523 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119448.4 NP_195020.1 2 - 3 751 98
GenBank mRNA AF370160.1 - 2 - 3 463 98
AF424619.1 - 2 - 3 461 98
AY059104.1 - 2 - 3 389 98
AY133531.1 - 2 - 3 389 98

View Full Product Details