Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEA4
Entrez Gene ID
816479
Gene Name
K+ efflux antiporter 4
Gene Aliases
AT2G19600, ATKEA4, F3P11.20, F3P11_20, K+ efflux antiporter 4
Chromosome Location
Chr.2: 8478970 - 8483854 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179659.2 NP_849990.1 19 - 20 1997 91

View Full Product Details