Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABG3c
Entrez Gene ID
820855
Gene Name
RAB GTPase homolog G3C
Gene Aliases
AT3G16100, ATRAB7D, ATRABG3C, RAB GTPase homolog G3C
Chromosome Location
Chr.3: 5459162 - 5460816 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112480.4 NP_188231.1 5 - 6 516 98
GenBank mRNA AB071848.1 - 5 - 6 408 98
AK227936.1 - 5 - 6 504 98
AY084691.1 - 5 - 6 510 98
BT006376.1 - 5 - 6 408 98

View Full Product Details