Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECT6
Entrez Gene ID
820996
Gene Name
evolutionarily conserved C-terminal region 6
Gene Aliases
AT3G17330, evolutionarily conserved C-terminal region 6
Chromosome Location
Chr.3: 5916940 - 5920281 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338284.1 NP_001327579.1 2 - 3 171 74
NM_112611.3 NP_188359.2 2 - 3 467 74
GenBank mRNA BT025338.1 - 1 - 2 70 74

View Full Product Details