Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALMT9
Entrez Gene ID
821373
Gene Name
aluminum-activated malate transporter 9
Gene Aliases
AT3G18440, AtALMT9, aluminum-activated malate transporter 9
Chromosome Location
Chr.3: 6328027 - 6330816 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112729.4 NP_188473.1 3 - 4 965 102
GenBank mRNA AY099663.1 - 3 - 4 846 102
BT000258.1 - 3 - 4 811 102

View Full Product Details