Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPC
Entrez Gene ID
821469
Gene Name
maternally expressed pab C-terminal
Gene Aliases
AT3G19350, maternally expressed pab C-terminal
Chromosome Location
Chr.3: 6705474 - 6706127 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112822.3 NP_188566.1 1 - 2 153 97
GenBank mRNA BT024667.1 - 1 - 2 101 97

View Full Product Details