Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFH12
Entrez Gene ID
829801
Gene Name
SEC14-like 12
Gene Aliases
AP22.26, AP22_26, AT4G36490, ATSFH12, SEC14-LIKE 12, SEC14-like 12
Chromosome Location
Chr.4: 17221885 - 17225088 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119812.4 NP_568006.1 7 - 8 1147 80
GenBank mRNA AY050419.1 - 7 - 8 1041 80
BT000834.1 - 7 - 8 933 80

View Full Product Details