Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CKL13
Entrez Gene ID
839520
Gene Name
casein kinase like 13
Gene Aliases
AT1G04440, F19P19.10, F19P19_10, casein kinase like 13
Chromosome Location
Chr.1: 1202160 - 1206009 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100324.3 NP_171939.1 12 - 13 1642 77
GenBank mRNA AY943851.1 - 12 - 13 987 77
BT015792.1 - 12 - 13 987 77
BT021997.1 - 12 - 13 1076 77

View Full Product Details