Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATX2
- Entrez Gene ID
-
837093
- Gene Name
- trithorax-like protein 2
- Gene Aliases
-
AT1G05830, SDG30, SET DOMAIN PROTEIN 30, T20M3.10, T20M3_10, trithorax-like protein 2
- Chromosome Location
-
Chr.1: 1754130 - 1761909 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100464.7 |
NP_172074.6 |
25 - 26 |
3627 |
146 |
View Full Product Details