Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- OFP4
- Entrez Gene ID
-
837202
- Gene Name
- ovate family protein 4
- Gene Aliases
-
ARABIDOPSIS THALIANA OVATE FAMILY PROTEIN 4, AT1G06920, ATOFP4, F4H5.2, F4H5_2, ovate family protein 4
- Chromosome Location
-
Chr.1: 2124676 - 2125902 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100566.2 |
NP_172174.1 |
1 - 1 |
998 |
134 |
View Full Product Details