Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLT1
Entrez Gene ID
821632
Gene Name
Integrase-type DNA-binding superfamily protein
Gene Aliases
AT3G20840, PLETHORA 1
Chromosome Location
Chr.3: 7300444 - 7303693 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112975.3 NP_188720.2 8 - 9 1264 108
GenBank mRNA AY506549.1 - 8 - 9 1033 108

View Full Product Details