Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIC
Entrez Gene ID
837394
Gene Name
Myosin family protein with Dil domain-containing protein
Gene Aliases
AT1G08730, ATXIC, F22O13.200, F22O13_200, MYOSIN XI C
Chromosome Location
Chr.1: 2779438 - 2788477 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331787.1 NP_001321098.1 33 - 34 4716 77
NM_100746.3 NP_172349.2 33 - 34 4215 77
GenBank mRNA LC177512.1 - 32 - 33 4191 77

View Full Product Details