The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_125559.3 | NP_200973.1 | 1 - 2 | 442 | 145 |
GenBank mRNA | BT010852.1 | - | 1 - 2 | 387 | 145 |
BT011324.1 | - | 1 - 2 | 368 | 145 |