Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMB3010
Entrez Gene ID
830900
Gene Name
Ribosomal protein S6e
Gene Aliases
AT5G10360, F12B17.290, F12B17_290, RPS6B, Ribosomal protein small subunit 6b, embryo defective 3010
Chromosome Location
Chr.5: 3258404 - 3260254 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203345.1 NP_001190274.1 3 - 4 221 68
NM_121074.5 NP_196598.1 3 - 4 241 68
GenBank mRNA AY045594.1 - 3 - 4 214 68
AY057634.1 - 3 - 4 214 68
AY093783.1 - 2 - 3 129 68

View Full Product Details