Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUX1
Entrez Gene ID
822350
Gene Name
plant UBX domain-containing protein 1
Gene Aliases
AT3G27310, K17E12.13, plant UBX domain-containing protein 1
Chromosome Location
Chr.3: 10087106 - 10089260 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113645.3 NP_566815.1 3 - 4 434 94
GenBank mRNA AK175284.1 - 3 - 4 263 94
AY085053.1 - 3 - 4 275 94
AY572781.1 - 3 - 4 251 94
BT024476.1 - 3 - 4 251 94

View Full Product Details