Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIR1
Entrez Gene ID
836395
Gene Name
SPFH/Band 7/PHB domain-containing membrane-associated protein family
Gene Aliases
AT5G62740, ATHIR1, AtHIR4, HIR4, HYPERSENSITIVE-INDUCED RESPONSE PROTEIN 1, MQB2.40, MQB2_40, hypersensitive induced reaction 4
Chromosome Location
Chr.5: 25200838 - 25202839 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125669.3 NP_201080.1 4 - 5 576 104
GenBank mRNA AY062850.1 - 4 - 5 467 104
AY086631.1 - 4 - 5 467 104
AY114572.1 - 3 - 4 404 104

View Full Product Details