Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUA
Entrez Gene ID
844268
Gene Name
nuclear pore anchor
Gene Aliases
AT1G79280, AtTPR, TRANSLOCATED PROMOTER REGION, YUP8H12R.12, YUP8H12R_12, nuclear pore anchor
Chromosome Location
Chr.1: 29818970 - 29833016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198506.1 NP_001185435.1 45 - 46 4875 94
NM_001198507.1 NP_001185436.1 46 - 47 4938 94
NM_106578.3 NP_178048.2 45 - 46 4834 94
GenBank mRNA EF426860.1 - 45 - 46 4665 94

View Full Product Details