The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_114531.3 | NP_190248.1 | 1 - 1 | 1322 | 173 |
GenBank mRNA | AK227686.1 | - | 1 - 1 | 1274 | 173 |