Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCA3
- Entrez Gene ID
-
823928
- Gene Name
- ABC2 homolog 2
- Gene Aliases
-
A. THALIANA ABC2 HOMOLOG 2, ABC2 homolog 2, AT3G47740, ATATH2, ATH2, ATP-binding cassette A3
- Chromosome Location
-
Chr.3: 17600521 - 17605128 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114642.4 |
NP_190358.3 |
1 - 2 |
206 |
104 |
View Full Product Details