Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPT1
Entrez Gene ID
824039
Gene Name
serine palmitoyltransferase 1
Gene Aliases
AT3G48780, ATSPT1, SERINE PALMITOYLTRANSFERASE 1, serine palmitoyltransferase 1
Chromosome Location
Chr.3: 18089020 - 18093252 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339379.1 NP_001325849.1 6 - 7 976 75
NM_114737.5 NP_190447.1 6 - 7 976 75
GenBank mRNA AB074928.1 - 6 - 7 681 75
AY054489.1 - 6 - 7 887 75
AY059882.1 - 6 - 7 788 75
AY114662.1 - 6 - 7 650 75
BT006615.1 - 6 - 7 650 75

View Full Product Details