Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPS21
Entrez Gene ID
832461
Gene Name
terpene synthase 21
Gene Aliases
AT5G23960, ATTPS21, MZF18.16, MZF18_16, terpene synthase 21
Chromosome Location
Chr.5: 8092901 - 8095341 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122301.4 NP_197784.2 4 - 5 1031 67

View Full Product Details