Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KNAT3
Entrez Gene ID
832593
Gene Name
homeobox protein knotted-1-like 3
Gene Aliases
AT5G25220, F21J6.18, F21J6_18, KNAT3 HOMEOBOX PROTEIN, KNOTTED1-like homeobox gene 3
Chromosome Location
Chr.5: 8735944 - 8738601 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036861.1 NP_001031938.1 1 - 2 815 90
NM_122431.3 NP_197904.1 1 - 2 945 90
GenBank mRNA X92392.1 - - 853 90

View Full Product Details