Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL1
Entrez Gene ID
839024
Gene Name
ARG1-like 1
Gene Aliases
ARG1-like 1, AT1G24120, F3I6.4, F3I6_4
Chromosome Location
Chr.1: 8529064 - 8532262 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102258.6 NP_173822.2 2 - 3 575 66
GenBank mRNA AY074564.1 - 2 - 3 502 66
AY226825.1 - 2 - 3 507 66
BT025997.1 - 2 - 3 259 66

View Full Product Details