Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALT1
Entrez Gene ID
839224
Gene Name
galactosyltransferase1
Gene Aliases
AT1G26810, T24P13.20, T24P13_20, galactosyltransferase1
Chromosome Location
Chr.1: 9286643 - 9290096 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332728.1 NP_001319087.1 7 - 8 1961 69
NM_102445.4 NP_174003.1 6 - 7 1921 69
GenBank mRNA AY133724.1 - 6 - 7 1901 69
EF428439.1 - 5 - 6 1614 69
KJ138985.1 - 5 - 6 1614 69

View Full Product Details