The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_102588.3 | NP_174144.1 | 1 - 2 | 935 | 70 |
GenBank mRNA | AK117664.1 | - | 1 - 2 | 762 | 70 |
BT005504.1 | - | 1 - 2 | 710 | 70 |