Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDA7
Entrez Gene ID
833525
Gene Name
histone deacetylase7
Gene Aliases
AT5G35600, AtHDA7, K2K18.5, K2K18_5, histone deacetylase7
Chromosome Location
Chr.5: 13770121 - 13771712 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122951.1 NP_198410.1 3 - 4 1101 91
GenBank mRNA AF510166.1 - 3 - 4 1101 91
DQ447001.1 - 3 - 4 1101 91

View Full Product Details