The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001344319.1 | NP_001331518.1 | 1 - 2 | 233 | 86 |
NM_123392.2 | NP_198844.1 | 1 - 2 | 281 | 86 | |
GenBank mRNA | AF214106.1 | - | 1 - 2 | 233 | 86 |
BT030457.1 | - | 1 - 2 | 233 | 86 |