Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERA1
Entrez Gene ID
834026
Gene Name
Prenyltransferase family protein
Gene Aliases
ARABIDOPSIS THALIANA FARNESYL TRANSFERASE BETA SUBUNIT, AT5G40280, ATFTB, ENHANCED RESPONSE TO ABA 1, FARNESYL TRANSFERASE BETA SUBUNIT, MSN9.180, MSN9_180, WIG, WIGGUM
Chromosome Location
Chr.5: 16101343 - 16104962 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344319.1 NP_001331518.1 4 - 5 460 75
NM_123392.2 NP_198844.1 4 - 5 508 75
GenBank mRNA AF214106.1 - 4 - 5 460 75
BT030457.1 - 4 - 5 460 75

View Full Product Details