Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPM1
Entrez Gene ID
834085
Gene Name
urophorphyrin methylase 1
Gene Aliases
AT5G40850, MHK7.8, MHK7_8, urophorphyrin methylase 1
Chromosome Location
Chr.5: 16365900 - 16369316 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203515.1 NP_001190444.1 3 - 4 1033 87
NM_001344363.1 NP_001331463.1 3 - 4 1033 87
NM_123450.5 NP_198901.1 3 - 4 1486 87
GenBank mRNA AF462833.1 - 3 - 4 872 87
AY058093.1 - 3 - 4 868 87
AY079029.1 - 3 - 4 872 87
AY142030.1 - 2 - 3 612 87
L47479.1 - 3 - 4 726 87

View Full Product Details