Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCA3;1
Entrez Gene ID
834324
Gene Name
Cyclin A3;1
Gene Aliases
AT5G43080, Cyclin A3;1, MMG4.10, MMG4_10
Chromosome Location
Chr.5: 17292952 - 17295024 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344470.1 NP_001331849.1 2 - 3 756 153
NM_123674.2 NP_199122.1 2 - 3 798 153

View Full Product Details