Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGLP2
Entrez Gene ID
834827
Gene Name
2-phosphoglycolate phosphatase 2
Gene Aliases
2-phosphoglycolate phosphatase 2, AT5G47760, ATPGLP2, ATPK5, MCA23.8, MCA23_8
Chromosome Location
Chr.5: 19342405 - 19345127 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001344745.1 NP_001332593.1 8 - 9 930 108
NM_124150.5 NP_199587.1 9 - 10 847 108
GenBank mRNA AK118640.1 - 9 - 10 714 108
BT005294.1 - 9 - 10 699 108

View Full Product Details